EW-48708-20
Handheld, lightweight, portable Digital Microscope, 1.3 megapixels, 10x to 200x magnification, USB connection
![]() |
Active Pixel Technology produces sharp images and accurate color reproduction |
![]() |
Includes software for image/video capture and measurement |
| Use these digital microscopes for science and engineering work, assembly and quality control, detailed repair, forensics, and more. Six LEDs illuminate your object—turn the LEDs on or off and adjust the brightness using the control wheel on the body. For versions with the 10x to 200x magnification range, magnification automatically adjusts based on the distance from the object. Includes a basic stand with adjustable height; order optional boom stand for greater flexibility. Capture images and videos through use of the included software (requires Windows® XP, SP2, or Vista). The software also includes a measurement function, brightness and contrast controls, digital zoom, and date and time record.What’s Included: software and microscope stand. |
|
Specifications
| Magnification | 10x to 200x | |
| Image sensor | inches | 1/4″ CMOS |
| Cable(s) | USB 2.0 | |
| Resolution | 1,300,000 pixels, 1280 x 1024 | |
| Light source | six LEDs | |
Related Articles
Cole-Parmer Introduces Enhancements to Life Science Offering
Sterilized Disposable Bioprocess Consumables
Basics of Cartridge Filtration
More Related TechInfo Articles