Learn how to Freeze Dry your dead cat
April 29, 2010
Foreword
This booklet has been developed to serve as a basic guide to the freeze drying process. The information presented is generic in nature and is the result of research and experience by Labconco personnel and users of freeze drying equipment. It is our intention to provide a non-biased review of preparation techniques and freeze drying methods. The purpose of this booklet is to help you make an informed choice of equipment for your laboratory applications.
Our Method
We begin our discussion of freeze drying for the laboratory by examining the three steps in the process: prefreezing, primary drying and secondary drying. Next, we examine a typical freeze drying cycle and the methods available to facilitate the freeze drying process using equipment designed for use by laboratories. Finally, suggestions to optimize successful results are discussed, including determination of end point, contamination, backfilling of dried samples and product stability. A glossary of terms used throughout this booklet to explain the freeze drying process follows the text, along with a bibliography.
Introduction
Freeze drying has been used in a number of applications for many years, most commonly in the food and pharmaceutical industries. There are, however, many other uses for the process including the stabilization of living materials such as microbial cultures, preservation of whole animal specimens for museum display, restoration of books and other items damaged by water, and the concentration and recovery of reaction products.
Specialized equipment is required to create the conditions conducive to the freeze drying process. This equipment is currently available and can accommodate freeze drying of materials from laboratory scale projects to industrial production.
Freeze drying involves the removal of water or other solvent from a frozen product by a process called sublimation. Sublimation occurs when a frozen liquid goes directly to the gaseous state without passing through the liquid phase. In contrast, drying at ambient temperatures from the liquid phase usually results in changes in the product, and may be suitable only for some materials. However, in freeze drying, the material does not go through the liquid phase, and it allows the preparation of a stable product that is easy to use and aesthetic in appearance.
The advantages of freeze drying are obvious. Properly freeze dried products do not need refrigeration, and can be stored at ambient temperatures. Because the cost of the specialized equipment required for freeze drying can be substantial, the process may appear to be an expensive undertaking. However, savings realized by stabilizing an otherwise unstable product at ambient temperatures, thus eliminating the need for refrigeration, more than compensate for the investment in freeze drying equipment.
Principles of Freeze Drying
The freeze drying process consists of three stages: prefreezing, primary drying, and secondary drying.
Prefreezing: Since freeze drying is a change in state from the solid phase to the gaseous phase, material to be freeze dried must first be adequately prefrozen. The method of prefreezing and the final temperature of the frozen product can affect the ability to successfully freeze dry the material.
Rapid cooling results in small ice crystals, useful in preserving structures to be examined microscopically, but resulting in a product that is more difficult to freeze dry. Slower cooling results in larger ice crystals and less restrictive channels in the matrix during the drying process.
Products freeze in two ways, depending on the makeup of the product. The majority of products that are subjected to freeze drying consist primarily of water, the solvent, and the materials dissolved or suspended in the water, the solute. Most samples that are to be freeze dried are eutectics which are a mixture of substances that freeze at lower temperatures than the surrounding water. When the aqueous suspension is cooled, changes occur in the solute concentrations of the product matrix. And as cooling proceeds, the water is separated from the solutes as it changes to ice, creating more concentrated areas of solute. These pockets of concentrated materials have a lower freezing temperature than the water. Although a product may appear to be frozen because of all the ice present, in actuality it is not completely frozen until all of the solute in the suspension is frozen. The mixture of various concentration of solutes with the solvent constitutes the eutectic of the suspension. Only when all of the eutectic mixture is frozen is the suspension properly frozen. This is called the eutectic temperature.
It is very important in freeze drying to prefreeze the product to below the eutectic temperature before beginning the freeze drying process. Small pockets of unfrozen material remaining in the product expand and compromise the structural stability of the freeze dried product. The second type of frozen product is a suspension that undergoes glass formation during the freezing process. Instead of forming eutectics, the entire suspension becomes increasingly viscous as the temperature is lowered. Finally the product freezes at the glass transition point forming a vitreous solid. This type of product is extremely difficult to freeze dry.
Primary drying: Several factors can affect the ability to freeze dry a frozen suspension. While these factors can be discussed independently, it must be remembered that they interact in a dynamic system, and it is this delicate balance between these factors that results in a properly freeze dried product.
After prefreezing the product, conditions must be established in which ice can be removed from the frozen product via sublimation, resulting in a dry, structurally intact product. This requires very careful control of the two parameters, temperature and pressure, involved in the freeze drying system. The rate of sublimation of ice from a frozen product depends upon the difference in vapor pressure of the product compared to the vapor pressure of the ice collector. Molecules migrate from the higher pressure sample to a lower pressure area. Since vapor pressure is related to temperature, it is necessary that the product temperature is warmer than the cold trap (ice collector) temperature. It is extremely important that the temperature at which a product is freeze dried is balanced between the temperature that maintains the frozen integrity of the product and the temperature that maximizes the vapor pressure of the product. This balance is key to optimum drying. The typical phase diagram shown in Figure 1 illustrates this point. Most products are frozen well below their eutectic or glass transition point (Point A), and then the temperature is raised to just below this critical temperature (Point B) and they are subjected to a reduced pressure. At this point the freeze drying process is started.
Figure 1
Some products such as aqueous sucrose solutions can undergo structural changes during the drying process resulting in a phenomenon known as collapse. Although the product is frozen below its eutectic temperature, warming during the freeze drying process can affect the structure of the frozen matrix at the boundary of the drying front. This results in a collapse of the structural matrix. To prevent collapse of products containing sucrose, the product temperature must remain below a critical collapse temperature during primary drying. The collapse temperature for sucrose is -32° C.
No matter what type of freeze drying system is used, conditions must be created to encourage the free flow of water molecules from the product. Therefore, a vacuum pump is an essential component of a freeze drying system, and is used to lower the pressure of the environment around the product (to Point C). The other essential component is a collecting system, which is a cold trap used to collect the moisture that leaves the frozen product. The collector condenses out all condensable gases, i.e; the water molecules, and the vacuum pump removes all non-condensable gases.
It is important to understand that the vapor pressure of the product forces the sublimation of the water vapor molecules from the frozen product matrix to the collector. The molecules have a natural affinity to move toward the collector because its vapor pressure is lower than that of the product. Therefore, the collector temperature (Point D) must be significantly lower than the product temperature. As can be noted in Table 1, raising the product temperature has more effect on the vapor pressure differential than lowering the collector temperature.
Table 1
A third component essential in a freeze drying system is energy. Energy is supplied in the form of heat. Almost ten times as much energy is required to sublime a gram of water from the frozen to the gaseous state as is required to freeze a gram of water. Therefore, with all other conditions being adequate, heat must be applied to the product to encourage the removal of water in the form of vapor from the frozen product. The heat must be very carefully controlled, as applying more heat than the evaporative cooling in the system can remove warms the product above its eutectic or collapse temperature.
Heat can be applied by several means. One method is to apply heat directly through a thermal conductor shelf such as is used in tray drying. Another method is to use ambient heat as in manifold drying.
Secondary drying: After primary freeze drying is complete, and all ice has sublimed, bound moisture is still present in the product. The product appears dry, but the residual moisture content may be as high as 7-8%. Continued drying is necessary at the warmer temperature to reduce the residual moisture content to optimum values. This process is called isothermal desorption as the bound water is desorbed from the product.
Secondary drying is normally continued at a product temperature higher than ambient but compatible with the sensitivity of the product. All other conditions, such as pressure and collector temperature, remain the same. Because the process is desorptive, the vacuum should be as low as possible (no elevated pressure) and the collector temperature as cold as can be attained. Secondary drying is usually carried out for approximately 1/3 to 1/2 the time required for primary drying.
How Freeze Drying Works
Refer to the phase diagram (Figure 1) and a typical sublimation cycle (Figure 2). The product is first cooled to below its eutectic temperature (Point A). The collector is cooled to a temperature approximately 20° C cooler than the product temperature, generally around -50 to -80° C. The product should be freeze dried at a temperature slightly lower than its eutectic or collapse temperature (Point B) since the colder the product, the longer the time required to complete primary drying, and the colder the collector temperature required to adequately freeze dry the product.
After the product is adequately frozen and the collector temperature achieved, the system is evacuated using a vacuum pump (Point C). At this point, primary drying of the product begins and continues until the entire frozen matrix appears dry. Heat input to the product may be achieved by several means such as increasing the shelf temperature in the case of tray drying, or using a liquid bath for manifold drying. While the collector and vacuum pump create the conditions for allowing sublimation to occur, heat input is really the driving force behind the whole process.
Heat input to the sample can be enhanced by controlling the pressure in the system at some level above the ultimate capability of the vacuum pump. Some freeze dryers incorporate vacuum control systems that automatically regulate the pressure to the preset level. This allows additional gas molecules to reside in the system thereby improving the conduction of heat to the sample. This improves the sublimation rate, reducing process time and associated energy costs. Care must be taken to prevent the pressure within the system from exceeding the ice vapor pressure of the product or melting of the sample may occur.
Figure 2
Heat input to the product must be very carefully controlled especially during the early stages of drying. The configuration of the product container and the volume of the contained product can affect the amount of heat that can be applied. For small volumes of material, evaporative cooling compensates for high levels of heat and drying is accelerated.
The volume and configuration of the suspension to be freeze dried often determines how the material is freeze dried. For example, the greater the ratio of the surface area to the volume of the suspension, the faster drying occurs. This is because a greater area for the water molecules to leave the product exists compared to the distance they have to travel to reach the surface of the frozen matrix. Drying occurs from the top of the product and initially the removal of water molecules is efficient. However, as the drying front moves down through the product, drying becomes more and more difficult. The water molecules must now travel through the dried portions of the product which impedes their progress. As the drying front moves farther and farther down the matrix, the application of heat to the product becomes more important (Figure 3).
Figure 3
Shell freezing as a method of prefreezing the product can increase the surface area to volume ratio by spreading out the frozen product inside the vessel (Figure 4). Shell freezing is accomplished by rotating the vessel in a low temperature bath causing the product to freeze in a thin layer on the inside surface of the vessel. The thickness of the frozen suspension depends on the volume of the product in comparison to the size of the vessel. Shell freezing is primarily used in conjunction with manifold drying.
The vacuum system is very important during freeze drying because the pressure must be maintained at a low As drying proceeds product level to ensure adequate water vapor flow from the temperature remains below shelf temperature product to the collector. A pressure gauge (commonly called a vacuum gauge) is used to monitor the pressure in the system during the drying process. Pressure can be expressed in several different units which are compared in Table 2. Some gauges measure condensable gases, while others do not. Those gauges that do not measure the condensable gases give an indication of the total pressure in the system. Gauges that do sense the condensable gases indicate a change in pressure during drying. These sensors can be used as an indication of the rate of drying, as well as the endpoint of the drying process.
Figure 4
Table 2
Freeze Drying Methods
Three methods of freeze drying are commonly used: (1) manifold drying, (2) batch drying, and (3) bulk drying. Each method has a specific purpose, and the method used depends on the product and the final configuration desired.
Manifold Method. In the manifold method, flasks, ampules or vials are individually attached to the ports of a manifold or drying chamber. The product is either frozen in a freezer, by direct submersion in a low temperature bath, or by shell freezing, depending on the nature of the product and the volume to be freeze dried. The prefrozen product is quickly attached to the drying chamber or manifold to prevent warming. The vacuum must be created in the product container quickly, and the operator relies on evaporative cooling to maintain the low temperature of the product. This procedure can only be used for relatively small volumes and products with high eutectic and collapse temperatures.
Manifold drying has several advantages over batch tray drying. Since the vessels are attached to the manifold individually, each vial or flask has a direct path to the collector. This removes some of the competition for molecular space created in a batch system, and is most ideally realized in a cylindrical drying chamber where the distance from the collector to each product vessel is the same. In a “tee” manifold, the water molecules leaving the product in vessels farthest from the collector experience some traffic congestion as they travel past the ports of other vessels.
Heat input can be affected by simply exposing the vessels to ambient temperature or via a circulating bath. For some products, where precise temperature control is required, manifold drying may not be suitable.
Several vessels can be accommodated on a manifold system allowing drying of different products at the same time, in different sized vessels, with a variety of closure systems. Since the products and their volumes may differ, each vessel can be removed from the manifold separately as its drying is completed. The close proximity to the collector also creates an environment that maximizes drying efficiency.
Batch Method. In batch drying, large numbers of similar sized vessels containing like products are placed together in a tray dryer. The product is usually prefrozen on the shelf of the tray dryer. Precise control of the product temperature and the amount of heat applied to the product during drying can be maintained. Generally all vials in the batch are treated alike during the drying process, although some variation in the system can occur. Slight differences in heat input from the shelf can be experienced in different areas. Vials located in the front portion of the shelf may be radiantly heated through the clear door. These slight variations can result in small differences in residual moisture.
Batch drying allows closure of all vials in a lot at the same time, under the same atmospheric conditions. The vials can be stoppered in a vacuum, or after backfilling with inert gas. Stoppering of all vials at the same time ensures a uniform environment in each vial and uniform product stability during storage. Batch drying is used to prepare large numbers of ampules or vials of one product and is commonly used in the pharmaceutical industry.
Bulk Method. Bulk drying is generally carried out in a tray dryer like batch drying. However, the product is poured into a bulk pan and dried as a single unit. Although the product is spread throughout the entire surface area of the shelf and may be the same thickness as product dried in vials, the lack of empty spaces within the product mass changes the rate of heat input. The heat input is limited primarily to that provided by contact with the shelf as shown in Figure 5.
Bulk drying does not lend itself to sealing of product under controlled conditions as does manifold or batch drying. Usually the product is removed from the freeze dry system prior to closure, and then packaged in air tight containers. Bulk drying is generally reserved for stable products that are not highly sensitive to oxygen or moisture.
Figure 5
Determining Drying Endpoints
Several means can be used to determine the endpoint of primary drying. The drying boundary in batch drying containers has moved to the bottom of the product container and inspection reveals that no ice is visible in the product. No visible ice indicates only that drying at the edges of the container is complete and gives no indication of the conditions in the center of the product. An electronic vacuum gauge can be used to measure condensable gases in the system. When the pressure indicated by the electronic gauge reaches the minimum pressure attainable by the system, as measured by using a McLeod vacuum gauge or as determined previously, no more water vapor is leaving the product.
As the heat input to the product is increased, evaporative cooling keeps the product temperature well below the temperature of its surrounding atmosphere. When primary drying is complete, the product temperature rises to equal the temperature of its environment. In manifold systems and tray dryers with external collectors, the path to the collector can be shut off with a valve and the pressure above the product measured with a vacuum gauge. If drying is still occurring, the pressure in the system increases.
Contamination in a Freeze Dry System
Two types of contamination can occur in a freeze dry system. One results from freeze drying microorganisms and the other results from freeze drying corrosive materials.
The potential for contamination of a freeze drying system by microorganisms is real in any system where microorganisms are freeze dried without a protective barrier such as a bacteriological filter. Contamination is most evident in batch tray dryer systems where large numbers of vials are dried in a single chamber. Evidence for contamination can be found by sampling the surfaces of the vials, shelves and collector. The greatest degree of contamination is usually found on the vials and on the collector. Some vial contamination can be due to a bit of sloppiness in dispensing the material originally, but contamination on the collector is due to microorganisms traveling from the product to the collector through the vapor stream.
The potential for contamination must be considered every time microorganisms are freeze dried, and precautions must be taken in handling material after the freeze dry process is completed. Recognizing that the vials are potentially contaminated, the operator should remove the vials to a safe area such as a laminar flow hood for decontamination. Decontamination of the freeze dry system depends upon the type of freeze dry system used. Some tray dryer systems are designed for decontamination under pressure using ethylene oxide sterilization. Ethylene oxide is considered hazardous, corrosive and detrimental to rubber components. Its use should be carefully monitored. Coupled with the risk of contamination in a freeze dry system is the risk of cross contamination when freeze drying more than one product at time. It is not a good practice to mix microbiological products in a freeze dry system unless some type of bacteriological filter is used to prevent the microbial product from leaving the vial itself.
While freeze drying of corrosive materials does not necessarily present a risk to the operator, it does present a risk of damaging the freeze dry system itself. Freeze dry systems are designed using materials that resist corrosion and prevent the build up of corrosive materials. But care should be taken to clean the system thoroughly following each use to protect it from damage.
Backfilling
For many freeze dried products, the most ideal system of closure is while under vacuum. This provides an environment in which moisture and oxygen, both detrimental to the freeze dried material, are prevented from coming in contact with the product. In some cases, vacuum in a container may be less than ideal, especially when a syringe is used to recover the product, or when opening the vessel results in a rush of potentially contaminating air. In these cases, backfilling the product container with an inert gas such as argon or nitrogen is often beneficial. The inert gas must be ultrapure, containing no oxygen or moisture.
Backfilling of the product container is generally useful in a batch tray dryer type system. The backfilling should also be carried out through a bacteriological filter. It is important that the gas flow during backfilling be slow enough to allow cooling of the gas to prevent raising the collector temperature. Backfilling can be carried out to any desired pressure in those tray dryers that have internal stoppering capability, and the vials then stoppered at the desired pressure.
Stability of Freeze Dried Products
Several factors can affect the stability of freeze dried material. Two of the most important are moisture and oxygen.
All freeze dried products have a small amount of moisture remaining in them termed residual moisture. The amount of moisture remaining in the material depends on the nature of the product and the length of secondary drying. Residual moisture can be measured by several means: chemically, chromatographically, manometrically or gravimetrically. It is expressed as a weight percentage of the total weight of the dried product. Residual moisture values range from less than 1% to 3% for most products.
By their nature, freeze dried materials are hygroscopic and exposure to moisture during storage can destabilize the product. Packaging used for freeze dried materials must be impermeable to atmospheric moisture. Storing products in low humidity environments can reduce the risk of degradation by exposure to moisture. Oxygen is also detrimental to the stability of most freeze dried material so the packaging used must also be impermeable to air.
The detrimental effects of oxygen and moisture are temperature dependent. The higher the storage temperature, the faster a product degrades. Most freeze dried products can be maintained at refrigerator temperatures, i.e. 4-8° C. Placing freeze dried products at lower temperatures extends their shelf life. The shelf life of a freeze dried product can be predicted by measuring the rate of degradation of the product at an elevated temperature. This is called accelerated storage. By choosing the proper time and temperature relationships at elevated temperatures, the rate of product degradation can be predicted at lower storage temperatures.
________________________________
Glossary
  • Accelerated Storage: Exposure of freeze dried products to elevated temperatures to accelerate the degradation process that occurs during storage.
  • Batch Freeze Drying: Freeze drying multiple samples of the same product in similar sized vessels at the same time in a shelf tray dryer.
  • Bulk Freeze Drying: Freeze drying a large sample of a single product in one vessel such as the bulk drying pans designed for shelf tray dryers.
  • Collapse: A phenomenon causing collapse of the structural integrity of a freeze dried product due to too high a temperature at the drying front.
  • Collapse Temperature: The temperature above which collapse occurs. Collector: A cold trap designed to condense the water vapor flowing from a product undergoing freeze drying.
  • Internal Collector: A collector located in the same area as the product. All water vapor has a free path to the collector.
  • External Collector: A collector located outside the product area connected by a small port through which all water vapor must pass. Allows isolation of the product from the collector for drying end point determinations and easier defrosting.
  • Ethylene Oxide: A colorless, odorless gas used for gas sterilization of tray dryer systems.
  • Eutectics: Areas of solute concentration that freeze at a lower temperature than the surrounding water. Eutectics can occur at several different temperatures depending on the complexity of the product.
  • Eutectic Temperature: The temperature at which all areas of concentrated solute are frozen.
  • Evaporative Cooling: Cooling of a liquid at reduced pressures caused by loss of the latent heat of evaporation.
  • Freeze Drying: The process of drying a frozen product by creating conditions for sublimation of ice directly to water vapor.
  • Glass Transition Temperature: The temperature at which certain products go from a liquid to a vitreous solid without ice crystal formation.
  • Isothermal Desorption: The process of desorbing water from a freeze dried product by applying heat under vacuum.
  • Lyophilization: The freeze drying process.
  • Manifold Freeze Drying: A freeze drying process where each vessel is individually attached to a manifold port resulting in a direct path to the collector for each vessel.
  • Prefreezing: The process of cooling a product to below its eutectic temperature prior to freeze drying.
  • Pressure Gauge (Vacuum Gauge): An instrument used to measure very low pressures in a freeze drying system.
  • Thermocouple Gauge: A pressure gauge that measures only the condensable gases in the system. This gauge can be used as an indicator of drying end points.
  • McLeod Gauge: A mercury gauge used to measure total pressure in the system (i.e. condensable and non- condensable gases.)
  • Primary Drying: The process of removing all unbound water that has formed ice crystals in a product undergoing freeze drying.
  • Residual Moisture: The small amount of bound water that remains in a freeze dried product after primary drying. Residual moisture is expressed as the weight percentage of water remaining compared to the total weight of the dried product. The amount of residual moisture in a freeze dried product can be reduced during secondary drying.
  • Secondary Drying: The process of reducing the amount of bound water in a freeze dried product after primary drying is complete. During secondary drying, heat is applied to the product under very low pressures.
  • Shell Freezing: Freezing a product in a thin layer that coats the inside of the product container. Shell freezing is accomplished by swirling or rotating the product container in a low temperature bath.
  • Sublimation: The conversion of water from the solid state (ice) directly to the gaseous state (water vapor) without going through the liquid state.
  • Vapor Pressure: The pressure of the vapor in equilibrium with the sample.
Bibliography
  1. Barbaree, J.M. and A. Sanchez. 1982. Cross-contamination during lyophilization. Cryobiology 19:443-447.
  2. Barbaree, J.M., A. Sanchez and G.N. Sanden. 1985. Problems in freeze-drying: I. Stability in glass-sealed rubber stoppered vials. Developments in Industrial Microbiology 26:397-405.
  3. Barbaree, J.M., A. Sanchez and G.N. Sanden. 1985. Problems in freeze-drying: II. Cross-contamination during lyophilization. Developments in Industrial Microbiology 26:407-409.
  4. Flink, J.M. and Knudsen, H. 1983. An Introduction to Freeze Drying. Strandberg Bogtryk/Offset, Denmark.
  5. Flosdorf, E.W. 1949. Freeze-Drying. Reinhold Publishing Corporation, New York.
  6. Greiff, D. 1971. Protein structure and freeze-drying: the effects of residual moisture and gases. Cryobiology 8:145-152.
  7. Greiff, D. and W.A. Rightsel. 1965. An accelerated storage test for predicting the stability of suspensions of measles virus dried by sublimation in vacuum. Journal of Immunology 94:395-400.
  8. Greaves, R.I.N., J. Nagington, and T.D. Kellaway. 1963. Preservation of living cells by freezing and by drying. Federation Proceedings 22:90-93.
  9. Harris, R.J.C., Ed. 1954. Biological Applications of Freezing and Drying. Academic Press, New York.
  10. Heckly, R.J. 1961. Preservation of bacteria by lyophilization. Advances in Applied Microbiology 3:1-76.
  11. Heckly, R.J. 1985. Principles of preserving bacteria by freeze-drying. Developments in Industrial Microbiology 26:379-395.
  12. King, C.J. 1971. Freeze-Drying of Foods. CRC Press, Cleveland.
  13. May, M.C. E. Grim, R.M. Wheeler and J. West. 1982. Determination of residual moisture in freeze-dried viral vaccines: Karl Fischer, gravimetric, and thermogravimetric methodologies. Journal of Biological Standardization 10:249-259.
  14. Mellor, J.D. 1978. Fundamentals of Freeze-Drying. Academic Press, London.
  15. Nail, S.L. 1980. The effect of chamber pressure on heat transfer in the freeze-drying of parental solutions. Journal of the Parental Drug Association 34:358-368.
  16. Nicholson, A.E. 1977. Predicting stability of lyophilized products. Developments in Biological Standardization 36:69-75.
  17. Parkes, A.S., and A.U. Smith, Eds. 1960. Recent Research in Freezing and Freeze-Drying. Charles C. Thomas, Springfield.
  18. Rey, L.R., Ed. 1960. Traite de Lyophilization. Hermann, Paris.
  19. Rey, L.R., Ed. 1964. Aspects Theorique et Industriels de la Lyophilisation. Hermann, Paris.
  20. Rowe, T.W.G. 1970. Freeze-drying of biological materials: some physical and engineering aspects. Current Trends in Cryobiology: 61-138.
  21. Seligman, E.B. and J.F. Farber. 1971. Freeze-drying and residual moisture. Cryobiology 8:138-144.
An Industry Service Publication
reprinted with permission from Labconco Corporation
http://www.coleparmer.in/techinfo/techinfo.asp?htmlfile=guide-freezdrying.htm&ID=1034&referred_id=5618
How to pump beer, wine, soup, yogurt or boneless chicken meat
April 12, 2010
Processors are looking for efficient, CIP-able pumping systems at reasonable costs to perform more challenging tasks.
While standard centrifugal pumps are typically used for mixing, blending and transporting product from points A to B, moving product with minimal shearing requires a positive-displacement (PD) pump. PD pumps can move food and beverages with a minimum of shear because the product is moved, in the case of a rotary PD pump, within the rotor pockets from the inlet to the outlet.
Shear, the relative motion between adjacent layers of a moving liquid, affects different liquids in different manners. Paint thins in viscosity as it is stirred; but cornstarch and water thicken when sheared. With a low-shear pump, delicate soups can be transported from the kettle to the filler, keeping food particles intact, and yogurt can be moved in a dairy without subjecting it to shear that may cause it to separate later after packaging.
fex0410tu2.jpg<br>
Peristaltic pumps use rollers that pinch a tube, providing pumping action. Pumped product does not come in contact with the pump body, and the tube can be cleaned in place like piping. Source: Watson-Marlow.
Popular PD pumps for low-shear applications, according to Chuck Lewis, marketing specialist at Moyno Inc., include rotary types such as peristaltic, lobe, and eccentric disk pumps. In addition, diaphragm pumps belong to the reciprocating family, but like the three rotary types, are able to move product safely without shear. Other PD types include gear, progressive cavity, sinusoidal plate (sine) and rotary vane pumps. While you may want to leave the shearing to a controlled environment such as a Breddo Liwifier rather than a pump, for high shear applications, Lewis suggests using pumps that include grinders and other similar equipment meant to masticate product into smaller pieces that are then conveyed or pumped. He says the violent action of high-shear grinders is designed to help with the disposal process of many waste products of less value.
“If you want a pump for high-shear applications, the best pump technology would be centrifugal,” says Eric Nofziger, Cole-Parmer product manager. “There are specific designs that can accommodate the chopping and grinding of particulates in the system water,” he adds. The key item to take into consideration is the size of the suspended particles. Depending on the size constraint, the pump’s impeller can be either a traditional closed or an open one—and if open, the geometry is adjusted to take the size into consideration.
A gentler, kinder pump
Enlarge this picture
fex0410tu3_lrg.jpg<br>
Two pumps are compared for slip. The pump on the left has a tight performance band and shows a slip of 4.2 gpm, while the pump on the right has a wide or loose performance band and produces a slip of 28.2 gpm. Source: Pump Solutions Group.
When processors have a viscous product or a product that needs to be transferred gently, PD pumps are the pumps of choice, says Chuck Treutel, Watson-Marlow food & beverage division sales manager. Typically, PD pumps are used in applications where product integrity is paramount. They run at slower speeds and handle higher viscosities than centrifugal or rotary pumps. The most common type of PD pump found in the food industry is the rotary lobe or circumferential piston. These pumps have two shafts and two rotors that turn opposite each other where the meshing rotors create a vacuum at the inlet pulling product in, carrying the product through and discharging it.
Lobe pumps are not great for metering duties because fluid slips between the lobes and the case, explains Treutel. While lobe pumps are low-shear devices, peristaltic pumps are better PD devices as they use a pinched-tube principle to move product, resulting in lower slippage and shear. Another option, says Treutel, is the sinusoidal pump, which has a single sinusoidal rotor that’s capable of powerful suction with low shear, low pulsation and gentle handling.
Treutel points to a leading poultry processor that used two sinusoidal pumps in its marinated boneless breast application, replacing vacuum and air pumps that were previously used to transfer the product into a drop hopper to meter product onto a conveyor. The drop hopper was also replaced, and the sinusoidal pumps were set up to meter boneless products through end-user-supplied spreader horns. Two pumps and two horns were arranged to feed two 16-in.-wide, half-inch sheets of boneless breast meat onto the conveyor, creating an almost-labor-free roasting line.
While perhaps similar in concept to a peristaltic or sinusoidal pump, Mouvex’s eccentric movement technology produces a peristaltic effect with a disk moving in an eccentric (non-rotating) motion inside an annular cylinder, says Wallace Wittkoff, Pump Solutions Group global hygienic director. Because the pumping elements don’t rotate, but instead move in an eccentric motion, there is no need for a mechanical seal. A rubber boot or metal bellows is used to accommodate this eccentric motion. For very low- or almost no-shear applications, the eccentric pump and an old standby, the air-operated diaphragm pump, both are good choices for delicate food products, adds Wittkoff.
Shear, slip, efficiency and saving energy
fex0410tu4.jpg<br>
This winery pump with a progressing cavity design moves up to 20 tons of must or whole clusters of grapes per hour and includes a built-in auger-feed mechanism. Source: Moyno.
“Since the initial price of a pump is only about 5% of its life cycle costs, I advise customers to look at the total cost of ownership of the equipment,” says Sam Raimond, Fristam Pumps customer service supervisor. “This includes maintenance costs (reliability, CIP-ability), more efficient motors and pump efficiency (energy costs),” he adds.
Processors want to save energy, and one way is to use efficient pumps. Shear and slip are inter-related and can affect pump efficiency. Pumps that are best for low-shear applications typically run at slow speeds, have small-diameter rotating parts, and produce minimal internal slip, says Mike Dillon, president of seepex Inc. By definition, PD pumps have the lowest shear rates, he adds.
In pump design, the higher the pump efficiency, the tighter the performance and the greater the energy savings. “When looking at centrifugal pumps, you want a design that utilizes most of its energy for moving liquid instead of being converted into thermal energy,” says Jim LeClair, SPX Flow Technology global product manager. In the design of rotary PD pumps, the minimization of slip within the rotor and case design maximizes the pump’s efficiency, he adds. Finding the optimum pump size based on pumping efficiency is important in maximizing energy savings.
For PD pumps, it’s assumed that no slip equals tight performance. But in reality, most rotary PD pumps do not attain true positive displacement because of slip. The actual flow produced is subject to product viscosity, back pressure, temperature (large clearances are needed) and wear, which is the most troubling, says Wittkoff. Since the Mouvex eccentric technology has very low slip, it can compress air, thus purging much of the product from the line when the supply tank runs dry, Wittkoff adds.
Fewer moving parts in the design of a pump can save energy. Sinusoidal pumps have only one set of bearings, one shaft and one rotor, compared to two of everything for the rotary lobe design, says Treutel.
“The result is less energy required to drive our pumps. Also peristaltic pumps are unique [because] they are 100% volumetrically efficient. This means zero slip,” he adds. No slip means very low shear is imparted on the product, and if the product is not slipping back from the discharge to the inlet, it means the product doesn’t have to be pumped twice, saving time and energy.
While the physical appearance of PD pumps has changed little over the years, better materials and improved technology have afforded internal pump changes that save energy, says Lewis. Now smaller drive ends can handle larger elements than in the past, which is both a cost and energy saver. Still the best way to save energy is to apply the right pump to the application. The correctly-sized pump can overcome friction loss, avoid excessive back pressure and still handle the upset condition.
“CIP-ability”
fex0410tu5.jpg<br>
For maximum overall efficiency, a rotary lobe pump can be teamed up with a variable-frequency drive to find the optimum spot on the pump’s performance curves. Source: Cole-Parmer.
The term CIP-ability implies the ability for a device to be compatible with clean-in-place methods—to be able to withstand harsh chemicals and extreme wash temperatures. But several criteria raise concern for processors. For example, how quickly and easily can a CIP-able pump be taken apart to verify cleaning? How can a CIP process in a pump be validated? Do seals and pumps exist that can withstand the CIP process? What approvals show that a pump is CIP-able?
“It seems as though the most desirable trait is ease and simplicity to manually clean,” says Dillon. Even though CIP has gained acceptance, many plants still disassemble pumps and piping for frequent inspection, he adds. This explains the persistence of lobe and centrifugal pumps in some plants when other designs are less expensive, are more CIP-able, have lower shear, and are more efficient, says Dillon.
Lewis says his company’s progressive cavity pumps take about an extra 5 to 10 minutes to disassemble and reassemble. The connections are made through a tri-clamp system that makes disassembly easy. More importantly, he adds, the pump should have regulatory approvals in place such as 3-A Sanitary Standards Inc., which indicate the pump has been rigorously tested to meet sanitary conditions.
Quick disassembly and repairs to CIP-able pumps depend on the design from the supplier, says Nofziger. Some designs have a wing-nut-type release bolt to remove the cover plate, while others require more extensive tools. As for repairs, front seals are easier to replace than rear seals, and working in the gearbox can be problematic, he adds. Gear-box repairs on lobe pumps can be tricky because of the timing issues. And with lobe pumps, Nofziger recommends not turning them on until the head temperature has stabilized to the fluid or product. Otherwise spalling could occur inside, causing slippage.
“Temperature variations, which are common in many food applications, can be very hard on pumps,” says Dillon. Dairy products are pumped cold, yet CIP or SIP (steam-in-place) is really hot. Tolerances can change and excessive wear can take place during CIP. Using thinner cross sections of temperature-sensitive materials—like elastomers—leads to less sensitivity to temperature fluctuations, he adds. seepex introduced a new line of 3-A pumps with thinner cross sections of elastomers in the stators to increase longevity in CIP applications.
Doug Silvey, CSI Designs solution expert, believes processors should spend the time normally spent on disassembling and reassembling pumps on validating the efficacy of CIP process on the pump. Adjusting the CIP time, cleaning chemicals, backpressure and pump speed when cleaning, and validating the pump is free of bacteria, eliminates repeated disassembly/re-assembly steps that can cause rotor alignment problems and damage.
The design of a CIP-able pump should meet all the requirements of the CIP definition as stated in the PMO and 3-A standards, says LeClair. When all equipment in the process line—including the pump—meets these standards, the processor can be assured that CIP will be effective for the entire line. Furthermore, if the pump has been tested to the European EHEDG standard, the processor can be assured that the pump will be clean according to the hygienic standards as outlined in the European Union, he adds.
One thing to keep in mind is that pump costs can be much higher if they have to comply with certain standards like 3-A.
“When a pump is rated 3-A, the pump manufacturer not only ensures the correct material of construction [is used], but also pays special attention to O-ring design (how the pump is connected to the rest of the system) as well as making sure there are no sharp corners in the fluid path,” says Nofziger. This detail ensures that the pump can be cleaned to eliminate the chance of bacteria growth. Additional costs accumulate in the third-party verification step where a 3-A member visits the pump manufacturer’s plant and reviews the engineering drawings, he adds.
Seal or no seal
Seals can be a sticky issue, especially with harsh CIP fluids and temperatures. Lewis suggests it may not be that mechanical seals fail due to undergoing CIP. Rather, it could be that the type of seal is incorrect for the process. Double mechanical seals may need to be considered, or the use of a magnetic drive can replace mechanical seals.
The materials used in seals have improved and the standard material provided has been optimized to fit 80% of applications that are currently on the market, says LeClair. The materials most used for rotary seals are silicon carbide against a carbon face. As new elastomeric technologies have been introduced, the use of new compounds has helped seals hold up against many of the new cleaning agents used today as well as improved their heat resistance, he adds.
“There are many options available for pumps to withstand chemical attack, high temperature and high pressures,” says Raimond. “We offer perfluoroelastomers (ASTM designation: FFKM), which are designed to withstand high temperatures and are compatible with many aggressive chemicals. Hastelloy and AL6XN metals have high chemical resistance,” he adds.
Pump manufacturers have addressed seal problems with exotic materials and designs, seal flushing systems and recommended plant process changes, says Treutel. “But the best way of preventing seal problems, such as leaks and premature wear, is to eliminate the seal altogether.
“If a [processor] is having seal problems and cannot find a solution, try a peristaltic pump…they do not have product seals,” he advises.
In the case of peristaltic pumps, during CIP the rollers or shoes that compress the tube are retracted so the pump becomes nothing more than an extension of the piping, says Treutel. If the piping becomes clean during CIP, then the pump will as well because the product and CIP fluid is contained within the tube at all times. Although the pump has no seals, there is some mechanical wear on the tube itself, but replacing it is generally easy.
Hygienic diaphragm pumps such as Wilden’s typically don’t have a mechanical seal; their diaphragm serves as isolation between product and the outside environment. According to Wittkoff, his company’s 3-A and EHEDG-approved pumps have no counter-moving surfaces, so there is no issue with material buildup or abrasive wear.
Trends and challenges
Most processors ultimately want CIP-able PD pumps, and that has been a focus for many suppliers. In creating a CIP-able PD pump, Fristam designed it such that not only the pump remains in place during CIP, but also all of its parts—covers and rotors, says Raimond. The challenge was to make the pump without modifying or increasing its clearances to accommodate CIP.
Suppliers have also dealt with some interesting applications. For example, Lewis describes a sanitary pump system for cold sausage with a viscosity topping 1,000,000 cp. The pump had to be made of stainless steel and designed for quick disassembly. It has also been used in icing, batter and dough applications.
LeClair describes pumping a mixture of fruit skins and grain husks. The low moisture content of this product made for a very abrasive application that required a rubber lobe pump.
Nofziger suggests that a real challenge is educating the customer in properly specifying a pumping system. Choosing a pump that is operating too close to either end of its performance curve tends to happen when a processor is looking for a pump mostly with a price point in mind.
While it is important to find something that will work within the budget, it is also important to review the performance curve of the pump. If a pump costs a bit more and provides an operating point closer to the middle of the curve, it would be a better choice than one closer to the end of its curve. When choosing a pump with an operational point that is close to the end of its curve, the end user will also be spending more on the operation of the pump versus one that is in the middle of the curve (which should have a better efficiency). If the pump is operating 24/7, this operational inefficiency could add up to many times more than the cost of the original equipment.
For more information:
Chuck Lewis, Moyno Inc., 937-327-3111, chuck.lewis@robn.com
Eric Nofziger, Cole-Parmer, 800-323-4340, enofziger@coleparmer.com
Chuck Treutel, Watson-Marlow, 608-883-6851, chuck.treutel@wmpg.com
Wallace Wittkoff, Pump Solutions Group, 502-905-9169,
wallace.wittkoff@pumpsg.com
Jim LeClair, SPX Flow Technology, 262-728-4912, jim.leclair@apv.com
Mike Dillon, seepex Inc., 937-864-7150, mdillon@seepex.net
Sam Raimond, Fristam Pumps, 608-831-5001
Doug Silvey, CSI Designs, 800-654-5635, dougs@csidesigns.com
Wayne Labs, Senior Technical Editor
labsw@bnpmedia.com
Original article:  http://www.foodengineeringmag.com/Articles/Feature_Article/BNP_GUID_9-5-2006_A_10000000000000795034
To view Cole-Parmer’s pump supply:  http://www.coleparmer.com/catalog/product_index.asp?cls=47660&referred_id=5618
Lease your Lab Equipment from Cole-Parmer!
April 9, 2010
Finance your lab equipment from Cole-Parmer
Leasing available
The convenience of lease financing is now available for Cole-Parmer’s broad selection of fluid handling systems, laboratory equipment, and instrumentation.
Leasing through Cole-Parmerhas several benefits: 
  • Expanding your existing equipment budget
  • Conserving cash with 100% fixed monthly payments
  • Preserving current credit and bank lines
  • Easy upgrades to new equipment technologies
  • Tax benefit eligibility
  • A variety of end of term lease options
Program Overview
For any order of $3,000 or more (with 80% of the value from equipment), Cole-Parmer offers you a number of financing options including: 
  • Variety of purchase options such as Fair Market Value, Fixed 10% Purchase Option, or $1.00 Buyout
  • Leasing contract, Equipment Sale Agreements, or non-cancelable Rental contracts
  • Monthly customer payment options available
  • Program available for U.S. customers only.
For more information
contact one of our representatives today at 800-323-4340 extension 6055 available from 7:00 a.m. to 5:00 p.m.CST
Cole-Parmer makes the lease financing process easy!
  1. Contact a Cole-Parmer Representative today to receive a lease financing quote on your next equipment order.
  2. Complete a simple lease application via fax, e-mail, or e-form.
  3. Upon credit approval and signature of the lease financing documents, your equipment will be immediately shipped to you.
It’s that easy! 
Lease Forms
Lease Application e-form  Complete list of
Product Categories 
or e-mail us at
Leasing@coleparmer.com 
Full details:  http://www.coleparmer.com/catalog/leasing_program.asp?referred_id=5618
Get a handheld microscope for your computer for $199!
April 7, 2010
Imagine plugging a microscope into the USB port of your PC or laptop and viewing images with 10x to 200x or even 500x magnification. The latest models of handheld microscopes from Cole-Parmer offer just this portability, convenience, and versatility. The most recent model also contains a polarizer for reducing glare on reflective items, such as metal surfaces.
Each of the Digital USB Microscopes produces crisp, sharp images and accurate color reproduction through the use of Active Pixel Technology. A simple press of the shutter trigger quickly captures the high-resolution magnified image. Six built-in lights with adjustable brightness ensure proper illumination. Software for image and video capture, measurement, brightness and contrast controls, digital zoom, and date-and-time record is included (requires Windows® XP, SP2, or Vista).
Customers working in laboratories, research, and quality control cite functionality, easy set-up, and economical price as significant reasons to recommend the devices. Scientists, engineers, forensics specialists, and technicians may also rely on the handheld microscopes to perform detailed repair, assembly, or inspection.
EW-48708-20
Handheld, lightweight, portable Digital Microscope, 1.3 megapixels, 10x to 200x magnification, USB connection
Digital Microscope
Active Pixel Technology produces sharp images and accurate color reproduction
Includes software for image/video capture and measurement
Use these digital microscopes for science and engineering work, assembly and quality control, detailed repair, forensics, and more. Six LEDs illuminate your object—turn the LEDs on or off and adjust the brightness using the control wheel on the body. For versions with the 10x to 200x magnification range, magnification automatically adjusts based on the distance from the object. Includes a basic stand with adjustable height; order optional boom stand for greater flexibility.
Capture images and videos through use of the included software (requires Windows® XP, SP2, or Vista). The software also includes a measurement function, brightness and contrast controls, digital zoom, and date and time record.What’s Included: software and microscope stand.
Specifications
Magnification 10x to 200x
Image sensor inches 1/4″ CMOS
Cable(s) USB 2.0
Resolution 1,300,000 pixels, 1280 x 1024
Light source six LEDs
Related Articles
Cole-Parmer Introduces Enhancements to Life Science Offering
Sterilized Disposable Bioprocess Consumables
Basics of Cartridge Filtration
More Related TechInfo Articles
Get a FREE copy of the Cole-Parmer® Life Science Products & Solutions Catalog
April 5, 2010
Cole-Parmer® Life Science Products & Solutions Catalog — This 48-page catalog is targeted for Life Science applications such as cell biology, tissue and cell culture, electrophoresis, fermentation, genomics, histology, and microbiology. Features high-quality laboratory instrumentation including thermal cyclers, microscopes, balances, centrifuges, pH meters, pumps, and more.
Life Science Products & Solutions Catalog
http://www.coleparmer.com/requests/default.asp?referred_id=5618&sel=ZX
Follow
Get every new post delivered to your Inbox.
Join 684 other followers