Summer pHun begins with proper pH
May 27, 2010
This weekend marks the unofficial start of summer. Millions will be heading to the swimming pool as public pools across the country officially open for business. Before jumping into any pool, keep in mind the importance of proper pH levels in the water.  
Oakton® Waterproof pHTestr™ 10 pH meter
Oakton® Waterproof pHTestr™ 10
Proper water pH is arguably one of the most critical aspects of maintaining correct swimming pool chemistry. Many articles describe how to properly maintain your pool chemistry and pH levels, so this post won’t detail how to manage your pool chemistry. Instead, think of this as a public service reminder to consider the importance of pool pH and take steps to check your swimming pool’s levels. Add chemicals as appropriate.To determine your pH levels, get a pH testing device from a local pool supply store or hardware store. Cole-Parmer also offers an option for private pool owners or large public swimming pools. Check out our line of pH testing meters.  
Our compact Oakton® Waterproof pHTestr 10  pH meters are great for outdoor use, including taking pH measurements in lakes, streams, and even swimming pools. These handheld pH meters are designed to float in water—and can be attached to a line running to the house for easy retrieval. The compact pH Testr 10 is even small enough to carry in your purse or pocket and take to a public pool or local beach to ensure that the water is safe. It is encased in Valox® sensor housing, which resists most chemicals. All waterproof pHTestrs also have replaceable sensors for easy maintenance.
Electrode sensors include a clog-resistant Kynar® junction and a recessed, glass pH bulb to protect against breakage. The keypad clicks to confirm operation, and allows access to a variety of functions. Calibrate up to three pH points with the auto-buffer recognition at both USA and NIST standard buffers. Additional features include automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.    
pHydron Ultrafine pH Paper Set Strips
pHydron Ultrafine pH Paper Strips
Another alternative for basic confirmations of pH, are pH indicators and pH paper strips from Cole-Parmer.
Let the pHun begin!    
We want to hear from you. Leave a comment and share how you plan to spend Memorial Day weekend. Are you headed to the beach or pool? Any plans for a cookout at the park? What is your ideal Memorial Day weekend activity? 
WARNING, DANGER, CAUTION, NOTICE, DUCK CROSSING.. Need a custom sign made? We do that.
May 27, 2010
Custom Signs
Custom signs for your unique needs
A sign needs to be quickly seen and easily understood to make the maximum impact. You know exactly what message needs to be conveyed by the signs in your facility. Now you can design your own customized signs by following these easy steps:
1. Choose the sign material that best meets your application.
  • Adhesive Vinyl—this flexible, 4-mil pressure-sensitive material is ideal for your low-cost, lightduty applications.
  • Plastic—this medium-duty, general-purpose, 0.055″-thick polyethylene material is perfect for indoor and sheltered outdoor locations.
  • Aluminum—this 0.040″-thick material offers the strength of metal but without rusting. The surface is overlaminated to protect from splashes and dirt for long-term outdoor exposure.
2. Select OSHA or ANSI header if required to designate the severity of the hazard.
  • Danger (red)
  • Caution (yellow)
  • Warning (orange)
  • Notice (blue)
  • Safety First (green)
    See Construction of a Safety Sign (pdf) for definitions of these hazards.
    3. Select from the industry standard sizes below or tell us what special size you need.
    4. We will work together to design a sign that best meets your unique needs.
    • Simply write or draw a concept, or use our easy-to-follow custom sign planning form on ColeParmer.com/customsigns.
    • Add words, colors, logos, symbols, images, and even photo-quality graphics!
    • We will lay out your message and develop the design, creating your custom sign for maximum effect. State-of-the-art printing produces your sign in the most accurate format.
    To create your custom signs
    with the help of our
    Customer Service Representatives,
    please call
    800-323-4340 ext. SIGN (7446).
  • Size Price per sign
    1 2 to 5 6 to 9 10+
    Adhesive vinyl
    5″ x 7″ $ 42.00 $ 24.00 $15.00 $ 8.00
    7″ x 10″   44.00   26.00   16.00  10.00
    10″ x 14″   47.00   29.00   20.00  13.00
    14″ x 20″   74.00   47.00   33.00  24.00
    20″ x 28″   90.00   62.00   48.00  38.00
    Custom size Call for a quote
    Plastic
    5″ x 7″   46.00   27.00   16.00   9.00
    7″ x 10″   48.00   29.00   19.00 12.00
    10″ x 14″   53.00   34.00   24.00 17.00
    14″ x 20″   84.00   56.00   41.00 30.00
    20″ x 28″ 111.00   77.00   58.00 44.00
    Custom size Call for a quote
    Aluminum
    5″ x 7″   46.00   27.00 18.00 12.00
    7″ x 10″   47.00   32.00 23.00 14.00
    10″ x 14″   56.00   41.00 32.00 27.00
    14″ x 20″   92.00   70.00 57.00 48.00
    20″ x 28″ 143.00 111.00 90.00 76.00
    Custom size Call for a quote
    Construction of a Safety Sign and Custom Sign order form
    Complete line of safety signs and labels
    For help from our
    Customer Service Representatives,
    call 800-323-4340
    ext. SIGN (7446).
    Unique problems require unique products.
    Let us help you get it right!
    View all the FREE
    Product Applications Support we offer.
    Cole-Parmer Products on Amazon.com
    May 21, 2010
    I’m proud to announce that as of 5/18, Cole-Parmer products will be available for purchase on Amazon.com and through Amazon.com affiliates. Over 41,000 Cole-Parmer products will be available on these sites to customers in the US. Amazon.com is the 7th highest trafficked Web site in the US and with 2009 revenue of 27B, they are the largest online retailer in the world.
    Customers will have the ability to purchase our products directly on Amazon’s Web sites using their Amazon.com account. Many of our products will be available, except those that are hazardous, require special documentation, have contract restrictions, etc.
    All purchases are backed by Amazon’s a-z guarantee.
    Cole-Parmer products on Amazon:
    http://www.amazon.com/gp/search/?brand=Cole-Parmer&ref_=bl_sr_industrial&node=16310161
    Cole-Parmer: http://www.coleparmer.com
    Count on Savings with Symmetry Balances
    May 21, 2010
    SYMMETRY CS COUNTING SCALE
    Industrial counting scales offer an accurate, efficient method for counting parts or items in your inventory.  Rather than counting items by hand, which is prone to error, use a counting scale to generate an accurate representation of the number of items in a parts bin.  What’s more, manually counting pieces by hand takes a considerable amount of time.  With the time saved in labor costs, a counting scale typically pays for itself after a few uses.
    Cole-Parmer is proud to announce the launch of our new industrial line of Symmetry CS Counting Scales.  Our counting scales expand upon our successful line of Symmetry balances.  Cole-Parmer’s Symmetry balances are designed to meet your budget as well as your every application need. They are not loaded with unnecessary and costly features your lab will never use—only those features you could never do without.
    Sweet Celebrations: Cole-Parmer India Moves Forward
    May 20, 2010
    In the midst of moving their office, growing their team, and keeping up with customer market demands, the Cole-Parmer business’s India team recently celebrated their fourth anniversary. Cole-Parmer’s Vice-President/General Manager Amit Agarwal traveled to the Mumbai office to join the Cole-Parmer staff for a reception and town hall meeting.  
    Collectively, the team has many milestones to recognize. Since its inception in 2006, sales have more than doubled. The team has expanded from 12 members to 30. An India-specific catalog was introduced within a year of formation and was followed by a Web site containing rupee pricing and in-stock status. Warehouse space has more than tripled. As the Cole-Parmer presence in India continues to swell, the leadership is implementing best practices in direct marketing and operations gleaned from the United Kingdom, United States, and Canadian teams.  
    “To enhance our exposure in the manufacturing hubs throughout India, we have hired key sales staff across the country,” said Jyothish Krishnan, Business Development Manager. “We have also augmented our product management resources to increase local sourcing and improve e-business capabilities. Our emphasis is on customer satisfaction. To this end, we have brought on board regional service professionals.”  
    Cole-Parmer India Moves Forward
    A new office location will enable the team to better serve customers through technical support and customer service.  
    As Cole-Parmer India keeps the momentum going, they savor the milestones they have already achieved. “We celebrated our recent anniversary with sweets,” noted Jyothish.  
    It seems fitting, as the future appears sweet for this growing hub of Cole-Parmer.
    Get your FREE copy of the Cole-Parmer® 2010 Lab Necessities Catalog!
    May 13, 2010
    Cole-Parmer 2010 Lab Necessities Catalog (LM, 99999-50)Cole-Parmer® 2010 Lab Necessities Catalog—Offering a vast assortment of labware, safety, and supplies, this 144-page catalog features best-selling laboratory products. Many are available at special volume savings—so you save more when you buy more. Plus, a variety of items are Guaranteed in Stock.
    Get your free catalog today:  http://www.coleparmer.com/requests/default.asp?sel=LM&referred_id=5618
    Win a Flip SlideHD Camcorder!
    May 10, 2010
    To enter, either place an order for $350 or more on coleparmer.com or send us a postcard (details below).  Orders place online are automatically entered in our drawing without any further action or information required.  This contest runs from 5/1/2010 thru 5/31/2010.
    win a Flip SlideHD Camcorder
    If you wish to enter without purchasing, you may do so by sending a postcard to: FLIP May 2010 Offer, c/o Cole-Parmer, eBusiness Department, 625 East Bunker Court, Vernon Hills, IL 60061 USA. On the postcard include: the date of mailing, your first and last name, company name, full shipping address, phone number and e-mail address.
     Flip SlideHD Camcorder features:
    • Record up to four hours of HD video
    • 3-inch wide touchscreen to show your favorite videos instantly
    • Store up to 12 hours of videos and photos in Space Saver format (using FlipShare software)
    • Connect the built-in USB arm to your PC or Mac to launch pre-loaded FlipShare software to organize, create and share your videos
    • Headphone jack (headphones not included) to enjoy your videos in any setting
    Full terms and conditions can be found here:  http://www.coleparmer.com/promotions/promotion_FLIPcamcorder2010.asp?referred_id=5618
    Get your free copy of our 2010 Water Quality catalog! [cue fireworks]
    May 7, 2010
    Get your free copy of our 2010 Water Quality catalog.
    This 96-page catalog features our newest and best-selling electrochemistry and water quality products, including a wide range of pH, conductivity, oxygen, turbidity, water sampling, and many more products.
    Request the new 2010 Cole-Parmer Water Quality today!
    http://www.coleparmer.com/requests/default.asp?sel=HA&referred_id=5618
    Washing lab glassware: Man vs. Machine
    May 5, 2010
    wandwash VS flaskscubber
    Glassware washing techniques vary from lab to lab and can depend on the soil type inside the glassware, types of glassware and technique required by the laboratory’s standard operating procedure.
    The three types of glassware washing we’ll look at are hand washing, residential dishwashers and laboratory glassware washers.
    Hand WashingHand washing requires an acid or base wash, rinse or soak which can be performed in plastic tubs. This method requires appropriate disposal of both acids and bases after washing is complete. Hot, soapy water can also effectively clean soiled glassware. A final rinse in purified water or tap water usually completes the process. In most cases, hand-washing water will reach 120ºF maximum, requiring sanitization or sterilization to be done in an autoclave. For drying, the glassware can be hung on a drying rack, placed in an oven, or dried manually. This process is time consuming and sometimes requires a dedicated person to hand wash the glassware.
    Automatic Residential DishwashersAutomatic residential dishwashers are another option sometimes employed when washing lab glassware. The initial investment is much less than an instrument designed for lab use and its features are not as durable or as flexible. The interior is generally plastic or stainless steel designed to handle basic food, soil and beverages, not chemicals and contaminants found in a laboratory. The baskets are designed for domestic plates, cups, bowls, glasses, pots and pans, rather than narrow neck glassware, culture tubes, flask, beakers, and other specialized glassware used daily in the laboratory. Basket inserts for lab utensils are also not available. Expect the manufacturer’s warranty to be void when used for purposes other than residential settings.
    Automatic Laboratory Glassware WashersAutomatic laboratory glassware washers are designed to clean all types of lab soil, from light to heavy, and offer features and accessories for a broad range of lab glassware and contaminants. A washer with a high heat option is best for cleaning heavy and difficult to remove soil such as waxes, oil or agar. For more soluble soil, high heat is not required. If flasks, graduated cylinders and other narrow-neck glassware are to be washed, a spindle rack option should be considered.
    The interior of a lab washer must be designed to prevent damage to the washer from residual exposure to basic laboratory chemicals. Stainless steel chambers withstand traces of the every day chemicals used in a laboratory. Internal components, including the detergent cup, seals, pumps and other plastic and rubber components, have been carefully chosen to withstand chemicals as well as the high heat conditions inside the washer.
    Another feature usually incorporated into a laboratory glassware washer is an optional purified water rinse. If pressurized purified water is not available, a separate pump may be required. Some washers include a separate pump to introduce purified water into the chamber, eliminating the need to buy it separately.
    HEPA filtered forced air drying, which traps dirt, lint and other particulate contaminants from the air, should be considered if your research is sensitive to this type of contamination. Also, an RS232 port, to communicate wash conditions to a printer or computer, is available on some models if validation is required.
    Pipette inserts allow the effective washing of pipettes in various sizes. The attachment forces water and detergent deep inside the pipettes assuring cleanliness. Other inserts for specific types of glassware such as culture tubes, media plates, DNA sequencing plates, and BOD bottles are readily available.
    Automatic washer detergents formulated specifically for laboratory washers can meet specific needs. There are a wide variety of detergents — phosphate-free, chlorine-free, surfactant-free and non-ionic or ionic. These detergents leave no residue on the glassware so they are safe for sensitive applications such as tissue culture or instrumental analysis. Some laboratory washers automatically dispense liquid detergent and weak acid neutralizing rinse solutions. This eliminates the need to manually fill the detergent cup and rinse dispenser before each use. Using a built-in pump, the washer automatically meters the precise amount of detergent and acidic rinse solution into the washer at the appropriate cycle.
    To determine the best washing system for your lab, an analysis should be done to calculate your potential daily cost of hand-washing vs. the cost of buying and using an automatic glassware washer. Information such as the amount of time spent per day hand-washing glassware, costs of labor, electricity, water, and detergent, types of glassware and how much of it — all should be included in the analysis. Once all the costs are calculated and compared, you can decide if the purchase of an automatic glassware washer provides a logical payback and consistent results. It’s working hard or working smart.
    Published with permission from Jenny Sprung, Senior Product Specialist, Labconco Corporation
    View our complete selection of laboratory glassware or laboratory glassware washers.
    Full article can be found at http://www.coleparmer.com/techinfo/techinfo.asp?htmlfile=glass-handVSmach.htm&ID=934&referred_id=5618
    Follow
    Get every new post delivered to your Inbox.
    Join 684 other followers