WARNING, DANGER, CAUTION, NOTICE, DUCK CROSSING.. Need a custom sign made? We do that.
Custom Signs
Custom signs for your unique needs
A sign needs to be quickly seen and easily understood to make the maximum impact. You know exactly what message needs to be conveyed by the signs in your facility. Now you can design your own customized signs by following these easy steps:
1. Choose the sign material that best meets your application.
  • Adhesive Vinyl—this flexible, 4-mil pressure-sensitive material is ideal for your low-cost, lightduty applications.
  • Plastic—this medium-duty, general-purpose, 0.055″-thick polyethylene material is perfect for indoor and sheltered outdoor locations.
  • Aluminum—this 0.040″-thick material offers the strength of metal but without rusting. The surface is overlaminated to protect from splashes and dirt for long-term outdoor exposure.
2. Select OSHA or ANSI header if required to designate the severity of the hazard.
  • Danger (red)
  • Caution (yellow)
  • Warning (orange)
  • Notice (blue)
  • Safety First (green)
    See Construction of a Safety Sign (pdf) for definitions of these hazards.
    3. Select from the industry standard sizes below or tell us what special size you need.
    4. We will work together to design a sign that best meets your unique needs.
    • Simply write or draw a concept, or use our easy-to-follow custom sign planning form on ColeParmer.com/customsigns.
    • Add words, colors, logos, symbols, images, and even photo-quality graphics!
    • We will lay out your message and develop the design, creating your custom sign for maximum effect. State-of-the-art printing produces your sign in the most accurate format.
    To create your custom signs
    with the help of our
    Customer Service Representatives,
    please call
    800-323-4340 ext. SIGN (7446).
  • Size Price per sign
    1 2 to 5 6 to 9 10+
    Adhesive vinyl
    5″ x 7″ $ 42.00 $ 24.00 $15.00 $ 8.00
    7″ x 10″   44.00   26.00   16.00  10.00
    10″ x 14″   47.00   29.00   20.00  13.00
    14″ x 20″   74.00   47.00   33.00  24.00
    20″ x 28″   90.00   62.00   48.00  38.00
    Custom size Call for a quote
    Plastic
    5″ x 7″   46.00   27.00   16.00   9.00
    7″ x 10″   48.00   29.00   19.00 12.00
    10″ x 14″   53.00   34.00   24.00 17.00
    14″ x 20″   84.00   56.00   41.00 30.00
    20″ x 28″ 111.00   77.00   58.00 44.00
    Custom size Call for a quote
    Aluminum
    5″ x 7″   46.00   27.00 18.00 12.00
    7″ x 10″   47.00   32.00 23.00 14.00
    10″ x 14″   56.00   41.00 32.00 27.00
    14″ x 20″   92.00   70.00 57.00 48.00
    20″ x 28″ 143.00 111.00 90.00 76.00
    Custom size Call for a quote
    Construction of a Safety Sign and Custom Sign order form
    Complete line of safety signs and labels
    For help from our
    Customer Service Representatives,
    call 800-323-4340
    ext. SIGN (7446).
    Unique problems require unique products.
    Let us help you get it right!
    View all the FREE
    Product Applications Support we offer.
    Leave a Reply
    Fill in your details below or click an icon to log in:
    WordPress.com Logo
    You are commenting using your WordPress.com account. Log Out / Change )
    Twitter picture
    You are commenting using your Twitter account. Log Out / Change )
    Facebook photo
    You are commenting using your Facebook account. Log Out / Change )
    Connecting to %s
    Follow
    Get every new post delivered to your Inbox.
    Join 1,219 other followers
    %d bloggers like this: