What’s Your Color? Portable Compact Color Meters Help You Find Out
Color Meters are ideal for checking painted surfaces, plastics, graphics, paper, textile, pharmaceuticals, and cosmetics. They also can be used in food and agriculture applications.
Cole-Parmer offers a Compact Color Meter that is lightweight and portable for use in the lab, plant, or field. Some models contain a sapphire window to lend extra protection to the color sensor.
The Compact Color Meters store readings in memory and recall readings on the display. Select color meters include software to enable users to manipulate and analyze data—download stored measurements through the USB port, create reports, upload standards, and create color libraries for automatic closest-color searching. The software includes formulas for CIELAB, Hunter Lab, CMC, LCh, and CIE94; it also calculates whiteness/yellowness indices, opacity, brightness, and delta E.
Leave a Reply
Fill in your details below or click an icon to log in:
WordPress.com Logo
You are commenting using your WordPress.com account. Log Out / Change )
Twitter picture
You are commenting using your Twitter account. Log Out / Change )
Facebook photo
You are commenting using your Facebook account. Log Out / Change )
Connecting to %s
Follow
Get every new post delivered to your Inbox.
Join 1,219 other followers
%d bloggers like this: