Stop by booth #669 on October 17-19 at the Los Angeles Convention Center. Cole-Parmer will be exhibiting at the Water Environment Federation Technical Exhibition and Conference (WEFTEC) and you can register for a chance to win an Apple® iPad®2!
You’ll also get to check out some of our latest water quality testing instruments and supplies. These include the new Thermo Scientific Orion Star A100 Meters—an excellent choice for environmental testing, water and wastewater treatment, quality control in food and beverage testing, and education. Affordable yet reliably effective, the meters feature a large LCD, simple button layout, and on-screen messages. Choose from units measuring pH, conductivity, or dissolved oxygen (DO).
The new Masterflex® L/S® Precision Variable-Speed Console Drives offer precise speed control and rpm display, which is ideal for metering applications. Confirm speed setting at a glance on the three-digit LED. With thereversible motor, you can purge tubing before or after pumping and pump fluid in either direction. The LED also indicates motor direction. Stack two or four pump heads for increased flow rates and multichannel applications.
The Oakton DO 2700 Benchtop Dissolved Oxygen Meter is designed for optimal performance and versatility. This advanced research grade meter has BOD capability. The meter measures dissolved oxygen in mg/L, ppm, or % saturation with simultaneous temperature display in °F or °C. Large LCD features annunciators to aid in meter use and troubleshooting. It is encased in a compact housing which takes up a minimum of bench space.