Industrial and Lab Safety: Monitoring Exposure to Toxic and Combustible Gases
Whether in industrial settings, manufacturing facilities, or as a matter of lab safety, gas monitors help protect workers by assessing their exposure to toxic and combustible gases. Air monitoring equipment is available in different varieties: an assessment must be conducted of potential hazards and then appropriate equipment chosen for use.
Here’s a description of various types of gas monitors:
GasAlertQuattro 4-Gas Detector, Rechargeable, %LEL, O2, H2S, CO
GasAlertQuattro 4-Gas Detector, Rechargeable, %LEL, O2, H2S, CO
Gas Detection Badgesthere are two types: badges with a color change indicating the presence of the gas in question; and badges that are sent to a lab after use for analysis. Badges with color change indicators are designed for a single chemical, but have the advantage of showing immediate results. The badges sent in for analysis can be used to sample a broader array of chemicals with higher accuracy at lower levels of concentration.
Gas Detection Tubesthere are two types: diffusion tubes and detector tubes that use a pump. Diffusion tubes allow gas to enter the tube through diffusion, react with the reagent, and produce a color change with exposure levels read from markings on the tube. These are used for time-weight average (TWA) calculations. Detector tubes are used with a hand pump for quick sampling of an area vs. longer term measurements for an individual worker.
Single- and Multi-Gas DetectorsCan be worn by a worker or handheld, giving an immediate reading of gas concentration. Visible and/or audible alarms alert you to high toxic gas concentrations, or low oxygen levels. Some models have data logging capabilities for later download to a computer for record-keeping purposes.
Photo-ionization detectors (PID) detect volatile organic compounds (VOCs). They are sensitive to a broad band of VOCs eliminating the need for multiple sensors. For best results, they should be calibrated using the target gas.
Leave a Reply
Fill in your details below or click an icon to log in:
WordPress.com Logo
You are commenting using your WordPress.com account. Log Out / Change )
Twitter picture
You are commenting using your Twitter account. Log Out / Change )
Facebook photo
You are commenting using your Facebook account. Log Out / Change )
Google+ photo
You are commenting using your Google+ account. Log Out / Change )
Connecting to %s