Selecting your Masterflex Tubing Pump System
There are three components in a complete Masterflex® Tubing Pump System. View below to see how these work together and what to consider for each:
Masterflex® Tubing Pump System
Masterflex® Tubing Pump System
Pump Head:
  • Flow rate—Different pump heads have different flow rate capabilities
  • Tubing change frequency—Some pump heads enable rapid tubing changes
  • Desired number of channels—Pump heads are stackable in most cases
  • Chemical compatibility—If chemicals spill, pump head materials become important
  • Accuracy—Standard pump heads offer the most accurate, repeatable flow rates
Tubing:
  • Flow rate—Tubing size directly affects flow rate
  • Chemical compatibility
  • Tubing life
  • Fluid viscosity, presence of solids, back pressure, or suction lift—High-performance tubing is best for these conditions
  • Fluid temperature
Drive:
  • Flow rate—The rpm of the drive directly correlates to flow rate
  • Fixed- or variable-speed—Variable-speed drives enable flow rate adjustment
  • Accuracy of flow rate or dispense volume—Many drives have digital speed control circuitry for highly accurate and repeatable flow rates
  • Remote control or computer interface—Some drives can connect to your process controller or can be operated by a foot switch
  • Environmental protection against hazards—Many drives are sealed against dust and water exposure
 The Complete Masterflex® System:
The Complete Masterflex® System
The Complete Masterflex® System
 See all Masterflex® Peristaltic Pumps, Pump Heads, and Tubing.
Leave a Reply
Fill in your details below or click an icon to log in:
WordPress.com Logo
You are commenting using your WordPress.com account. Log Out / Change )
Twitter picture
You are commenting using your Twitter account. Log Out / Change )
Facebook photo
You are commenting using your Facebook account. Log Out / Change )
Google+ photo
You are commenting using your Google+ account. Log Out / Change )
Connecting to %s
Follow
Get every new post delivered to your Inbox.
Join 1,403 other followers
%d bloggers like this: